Learn More
Medchemexpress LLC Darbufelone mesylate (CI-1004 mesylate) | 139340-56-0 | 99.7% | 428.57 | C19H28N2O5S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101438A100MG
Encompass_Preferred
Darbufelone mesylate is a small-molecule research reagent that inhibits cellular production of prostaglandin F2α (PGF2α) and leukotriene B4 (LTB4). It exhibits potent COX-2 (PGHS-2) inhibition and is supplied as a solid analytical standard for biochemical and pharmacological studies.
  • Potent inhibitor of PGHS-2 (COX-2); reported IC50 ~0.19 μM.
  • Dual inhibition of PGF2α and LTB4 production in cellular assays.
  • High purity suitable for analytical and pharmacological studies.
  • Supplied as a solid, white to light yellow material.
  • Molecular weight 428.57 and formula C19H28N2O5S2.
  • Includes downloadable COA, SDS, and data sheet for quality control.
Catalog No. 50-002-47883