Learn More
AdipoGen Darcin (mouse) (rec.) (untagged) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B00783010
Encompass_Preferred
Protein. Darcin (mouse) (aa 20-181) is untagged. Source: E. coli. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). Darcin (also called Mup20) belongs to the family of major urinary proteins (MUPs), a subfamily of proteins found in abundance in the urine. Like all MUPs, Darcin is a member of the lipocalin family of ligand-binding proteins, and preferentially binds the volatile pheromome (S)-2-sec-butyl-4,5-dihydrothiazole (SBT). Darcin is present in the urine of male and acts as a sex pheromone that elicits female sexual attraction to a male's urine scent. Recombinant Darcin shows similar activity compared to natural Darcin.
Catalog No. 50-205-3139
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.