Learn More
Apexbio Technology LLC Darifenacin HBr, 1g. Cas: 133099-07-7 MFCD: MFCD08141803. Selective M3 muscarinic receptor antagonist
SDP
Supplier:  Apexbio Technology LLC B16001000
Encompass_Preferred
DarifenacinHBr(UK88525) is a selective M3 muscarinic receptor antagonist with pKi of 8.9 [1]. Themuscarinic acetylcholine receptor, also known as cholinergic/acetylcholine receptor M3, or themuscarinic 3, is amuscarinic acetylcholine receptorencoded by the human geneCHRM3. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5272
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.