Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Darunavir
GREENER_CHOICE

Catalog No. 671010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-101-0 10 mg
67-105-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-101-0 Supplier Tocris Bioscience™ Supplier No. 6710/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent HIV protease inhibitor

Darunavir is a highly potent HIV protease inhibitor (IC50 = 3 - 6 nM, depending on laboratory HIV-1 strain). Displays activity in cells infected with clinical HIV-1 strains shown to have resistance to other protease inhibitors.

Tocris products are for biomedical research use only. They are not intended for human or veterinary use.

Specifications

Chemical Name or Material (3 R,3 AS,6 aR)-Hexahydrofuro[2,3-b]furan-3-yl N-[(1 S,2 R)-3-[[(4-aminophenyl)sulfonyl](2-methylpropyl)amino]-2-hydroxy-1-(phenylmethyl)propyl]carbamate
CAS 206361-99-1
Quantity 10 mg
Target HIV Protease Inhibitors
Molecular Formula C27H37N3O7S
Formula Weight 547.66
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.