Learn More
Cayman Chemical DastInIb N-oxIde 1mg
SDP
Supplier:  Cayman Chemical 278451 MG
Encompass_Preferred
A dopamine metabolite levels are increased 5- to 6-fold compared to wild-type in the nucleus accumbens striatum and prefrontal cortex of COMT knockout mice oxidized DOPAC binds to monomeric a-synuclein preventing fibrillation foot shock stress increases levels relative to control in the prefrontal cortex olfactory tubercles and VTA in rats
Catalog No. 50-293-7544
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.