Learn More
Anton Paar Autotap Dual station automated tap density analyzer
Supplier: Anton Paar 192980
The Dual-Autotap double station tapped density analyzer for powders is a simple to use, automated device which complies with a large number of internationally recognized standard methods. It provides a high level of test method control with a user-selectable, lockable number of taps. Tapping rate and drop height are fixed to provide consistent results from sample to sample. A large range of sample sizes can be accommodated with the use of a number of different graduated cylinder sizes. Tapped density measurement (sometimes referred to as tapped bulk density, or just tap density) is formalized in a number of international standards to which Autotap conforms.Includes two Graduated Cylinders 250 mL Tapping rate 260 per minuteTap counter 1 – 999,999Recommended Accessories: NC1818339, NC1564678
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.