Learn More
Cole-Parmer Data Acquisition Software with USB Cable; For 98766-89/90/93/94, 91210-03, 90080-04, and 98767-11
Supplier:  Cole-Parmer 3780413
Encompass_Preferred
Complete Data Acquisition Software enables you to display and store measurement readings on your PC from up to 8 devices at a time. Data capture runs in the background, enabling you to work on other programs, simultaneously. Set audible and visual notification alarms or immediately send an email if a over/under condition occurs. The software can be used to connect a meter directly to your PC via cable, or coupled with the Digi-Sense Wireless Radio-Signal Transceiver Kit (37804-12). Check software specification tables for meter compatibility, or contact our application specialists if you have a meter with RS-232 output not listed in the tables.
Catalog No. NC1910716
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.