Learn More
ChemDAQ™ Data Acquisition Hardware
Description
The ChemDAQ™ Data Acquisition System provides real-time data logging and centralized display of gas and vapor readings from up to 32 monitoring points simultaneously, giving you a comprehensive, facility-wide view of chemical exposure levels throughout your building. Designed for seamless integration, the system can transmit data to programmable logic controllers (PLCs) or facility management software, enabling your gas monitors to work as part of automated abatement systems or facility-wide alarm protocols.
Specifications
Specifications
| For Use With (Equipment) | ChemDAQ Gas Sensor |
| Type | Data Acquisition |
Purchase of this product requires an annual service contract for sensor calibration and exchange, which must be arranged prior to shipment. Customers in the food and beverage production and processing industry need to contact ChemDAQ directly to purchase this product. Contact your Fisher Scientific representative to confirm your needs and to request a quote.