Learn More
Thermco Products Data Logger Smart Probe
Catalog No. 04500303
Change view
Click to view available options
For Use With (Equipment):
Data Loggers
Catalog No. For Use With (Equipment)
04-500-303 Data Loggers
1 options
Catalog No. 04-500-303 Supplier Thermco Products Supplier No. VFCSP5
Add to Cart
Add to Cart
Calibrated SP-400-5 ft. Smart Temperature Probe - 5 ft. cable.
  • The 5 ft. Smart Probes for the VFC400-SP Data Logger deliver high-precision temperature monitoring for critical environments.
  • Hot-swappable design allows instant probe replacement without removing the logger, ensuring uninterrupted data collection.
  • Independently calibrated for superior accuracy of ±0.3°C from -40°C to +40°C.
  • Ideal for pharmaceuticals, food, and cold chain logistics requiring reliable temperature measurements.
  • Robust construction withstands extreme temperatures, high humidity, and intense pressure.
  • Each probe includes a NIST-traceable calibration certificate compliant with ISO/IEC 17025:2017.
  • Features a 3 in. stainless steel tip for durability and performance.
  • Plug-and-play functionality simplifies installation and use.
  • Backed by a 2-Years warranty to ensure compliance and peace of mind for vaccine and temperature-sensitive storage.
Specifications
Type Probe
For Use With (Equipment) Data Loggers