Learn More
Grainger HOBO Data Logger, 0 to 90% Relative Humidity, LCD Display, USB Interface, Internal Sensor
Supplier:  Grainger 38AX83
Encompass_Preferred
The HOBO Data Logger records 0-90% RH with ±2.5% accuracy. It displays real-time, min, max, and avg humidity on its LCD. It stores up to 21,500 readings, with adjustable logging intervals from 1 sec to 18 hrs. Powered by a replaceable lithium battery lasting up to a year, it features an audible alarm, is compact (3.5 x 2.25 x 0.9 inches), and includes mounting options. Compatible with HOBOware software for data analysis.
Catalog No. NC2092507
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.