Learn More
Roche Diagnostics Datalogic™ QD2590 Barcode Scanner
Catalog No. 21100539 Shop All Roche Diagnostics Products
Change view
Click to view available options
Dimensions (L x W x H):
7.87 x 5.91 x 3.15 in.
Catalog No. Dimensions (L x W x H)
21-100-539 7.87 x 5.91 x 3.15 in.
1 options
Catalog No. 21-100-539 Supplier Roche Diagnostics Supplier No. 09831053001
Add to Cart
Add to Cart
The Datalogic™ QD2590 barcode scanner is a trusted companion for use alongside Roche's Urisys 1100 urine analyzer.
The Datalogic™ QD2590 handheld barcode scanner (for use alongside Roche's Urisys 1100 urine analyzer and more) is versatile and easy to use, including the following features:
  • Omnidirectional reading for 1D, 2D, postal codes, stacked and composite bar codes, dot codes and Digital Watermark codes
  • Datalogic LED-based technology with extreme precision aiming system
  • Datalogic's 'Green Spot' technology for superior good-read feedback
  • Motionix patented motion-sensing technology for seamless reading mode transitions
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W x H) 7.87 x 5.91 x 3.15 in.
Type Barcode Scanner