Learn More
Thermo Scientific™ DataRAM™ Accessories
Catalog No. 19165934
Change view
Click to view available options
Description:
Omni-directional inlet sampling
Standard cartridge filter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
19-165-934 Omni-directional inlet sampling
19-165-936 Standard cartridge filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-165-934 Supplier Thermo Scientific™ Supplier No. DROSI
DataRAM™ Accessories have been discontinued. However, we continue to provide services and support for these products.
Accessory options include:
  • Omnidirectional sampling inlet
  • Temperature conditioning heater
  • In-line impactor head (10 and 2.5μm)
  • Impaction nozzle for 1μm
  • Ambient sampling inlet set
  • Isokinetic sampling set
  • Sampling dilution unit
  • Respirable cyclone precollector
  • Solar power supply
  • External DC cable assembly
  • Analog signal output cable
Recommended for:
  • Outdoor ambient sampling
  • PM-10, PM 2.5 and PM 1 testing
  • Humidity management 
  • Dilutions of high dust concentrations
TRUSTED_SUSTAINABILITY
Specifications
Description Omni-directional inlet sampling