Learn More
DATF1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300152AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Dio-1 (Death Inducer-Obliterator-1) is a putative transcription factor that contains two zinc finger motifs. Dio-1 translocates to the nucleus, and activates apoptosis during limb development. Programmed cell death, a highly regulated form of apoptosis, plays an important role in determining the amount of tissue, the shape, and the definition of each digit during limb development. Dio-1 expression is upregulated when an apoptotic signal is detected, and subsequently apoptosis is induced. This process is similar to the expression of NFkB and NGF in response to external signals. Dio-1 expression is suppressed by caspase inhibitors and Bcl-2 expression. This supports the theory that Dio-1 functions in the onset of programmed cell death.Specifications
| DATF1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9BTC0 | |
| DIDO1 | |
| Between 300 and 350 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| DIDO1 | |
| Death-inducer obliterator 1, DIO-1, hDido1, Death-associated transcription factor 1, DATF-1, DIDO1, C20orf158, DATF1, KIAA0333 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11083 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.