Learn More
Novus Biologicals™ Daudi Whole Cell Lysate

Specifications
Specifications
| Quantity | 500 μg |
| Purity | Multi-step |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | Daudi |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.