Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daunomycin, 95%, MP Biomedicals™

Catalog No. ICN15077710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15077710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15077710 Supplier MP Biomedicals, Inc Supplier No. 0215077710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sequence-specific double-stranded DNA intercalating agent used in antileukemic, photostability, and drug metabolism research.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23541-50-6
Molecular Formula C27H30ClNO10
Molecular Weight (g/mol) 563.984
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
Synonym Daunorubicin, Leukaemomycin C
PubChem CID 62770
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl

Specifications

Type Daunomycin
Physical Form Powder
Color Red
Quantity 10 mg
Assay Percent Range 0.95
Formula Weight 564
Grade Not Available
Chemical Name or Material Daunomycin, For HPLC analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.