CAS RN 23541-50-6
Daunomycin, 95%, MP Biomedicals™
CAS: 23541-50-6 Molecular Formula: C27H30ClNO10 Molecular Weight (g/mol): 563.984 InChI Key: GUGHGUXZJWAIAS-QQYBVWGSSA-N Synonym: Daunorubicin,Leukaemomycin C PubChem CID: 62770 IUPAC Name: (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride SMILES: CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl
Daunomycin hydrochloride, MP Biomedicals™
CAS: 23541-50-6 Synonym: Daunorubicin hydrochloride,Leukaemomycin C
Daunorubicin hydrochloride
CAS: 23541-50-6 Molecular Formula: C27H30ClNO10 Molecular Weight (g/mol): 563.984 MDL Number: MFCD04974507 InChI Key: GUGHGUXZJWAIAS-QQYBVWGSSA-N Synonym: daunorubicin hydrochloride,daunorubicin hcl,daunomycin hydrochloride,daunoblastina,rubidomycin hydrochloride,daunorubicinol hydrochloride,daunoblastin,rubomycin,ondena,daunomycin chlorohydrate PubChem CID: 62770 IUPAC Name: (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride SMILES: CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl
Daunorubicin Hydrochloride 98.0+%, TCI America™
CAS: 23541-50-6 Molecular Formula: C27H30ClNO10 Molecular Weight (g/mol): 563.984 MDL Number: MFCD04974507 InChI Key: GUGHGUXZJWAIAS-QQYBVWGSSA-N Synonym: daunorubicin hydrochloride,daunorubicin hcl,daunomycin hydrochloride,daunoblastina,rubidomycin hydrochloride,daunorubicinol hydrochloride,daunoblastin,rubomycin,ondena,daunomycin chlorohydrate PubChem CID: 62770 IUPAC Name: (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride SMILES: CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl
R&D Systems™ Daunorubicin hydrochloride
CAS: 23541-50-6 Molecular Formula: C27H30ClNO10