Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daunomycin hydrochloride, MP Biomedicals™

Catalog No. ICN15077750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15077750 50 mg
ICN15077701 1 mg
ICN15077705 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15077750 Supplier MP Biomedicals, Inc Supplier No. 0215077750
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Photostability, Antileukemic, and drug metabolism studies
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23541-50-6
Synonym Daunorubicin hydrochloride, Leukaemomycin C

Specifications

Type Daunomycin Hydrochloride
Physical Form Solid
Quantity 50 mg
Molecular Formula C27H30ClNO10
MDL Number MFCD04974507
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
Molecular Weight (g/mol) 563.984
PubChem CID 62770
Formula Weight 563.99
Grade Not Available
Chemical Name or Material Daunomycin hydrochloride
Show More Show Less
storageNote1 Protect from light.
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.