Learn More
Selleck Chemical LLC Rimonabant (SR141716) S3021-100mg
SDP
Supplier:  Selleck Chemical LLC S3021100MG
Encompass_Preferred
Rimonabant (SR141716) is a selective antagonist of CB1 with IC50 of 13 6 nM and EC50 of 17 3 nM in hCB1 transfected HEK 293 membrane Rimonabant is also a dual inhibitor of acyl CoA cholesterol acyltransferases(ACAT) 1 and 2 and inhibits mycobacterial MmpL3
Catalog No. 50-311-0740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.