Learn More
Medchemexpress LLC Daunorubicin hydrochloride | 23541-50-6 | MFCD04974507 | 99.7% | 563.98 g/mol | C27H30ClNO10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY130625MG
Encompass_Preferred
Daunorubicin hydrochloride is an anthracycline antibiotic and potent cytotoxic research compound that intercalates DNA and inhibits topoisomerase II. Provided as the hydrochloride salt for research use, it is commonly used in oncology and biochemical studies where controlled, small-quantity reagent handling and high-purity material are required.
  • Anthracycline antibiotic with cytotoxic activity.
  • Potent topoisomerase II inhibitor causing DNA intercalation.
  • Hydrochloride salt form for enhanced solubility and handling.
  • High purity suitable for analytical and biological assays.
  • Molecular weight 563.98 g/mol; formula C27H30ClNO10.
  • Supplied in small measured quantities for research workflows.
Catalog No. 50-002-63181