Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ DawnMist™ Deodorant

Catalog No. 22415797
Change view
Click to view available options
Volume (English):
1.6 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
22-415-797 1.6 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-415-797 Supplier Dukal™ Supplier No. SD175
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Stick Deodorant is alcohol free and fights odor with a clean fresh scent.

  • Fights odor
  • Clear plastic packaging for added security
  • Clean fresh scent
  • Alcohol free

Specifications

Product Type Stick Deodorant
Color Clear
For Use With (Application) Personal Care, Deodorant
Volume (English) 1.6 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.