Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daxx Mouse anti-Human, Unconjugated, Clone: DAXX5.14, MilliporeSigma™

Catalog No. MABE19111MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE19111MI 100 μg
MABE19112MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE19111MI Supplier MilliporeSigma Supplier No. MABE1911100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Daxx
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone DAXX5.14
Conjugate Unconjugated
Gene Accession No. NP_001341
Gene Alias Death domain-associated protein 6;hDaxx;ETS1-associated protein 1;EAP1;Fas death domain-associated protein
Gene Symbols DAXX, BING2, DAP6
Host Species Mouse
Immunogen His-tagged recombinant fragment corresponding to 318 amino acids from the C-terminal half of human Daxx.
Molecular Weight of Antigen 81.37 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone DAXX5.14 specifically detects human Daxx. It targets an epitope with in 40 amino acids from the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.