Learn More
Vector Laboratories DSL, Unconjugated, 5 mg FL-1031-5
SDP
Supplier:  Vector Laboratories FL10315
Encompass
Datura Stramonium Lectin (DSL), Unconjugated, 5 mg, contains two chains of 40 kDa and 46 kDa joined by disulfide bonds. This lectin is free of a reported 32 kDa contaminant protein. The carbohydrate binding site recognizes (beta-1,4) linked N-acetylglucosamine oligomers, preferring chitobiose or chitotriose over a single N-acetylglucosamine residue. This lectin binds well in the acidic pH range but its affinity decreases above pH 8.0. DSL also binds well to N-acetyllactosamine and oligomers containing repeating N-acetyllactosamine sequences. A branched pentasaccharide including two N-acetyllactosamine disaccharides linked to mannose (beta-1,6) and (beta-1,2) was reported to be the most potent inhibitor of agglutination. Inhibiting/Eluting Sugar: Chitin Hydrolysate.
Catalog No. 50-204-5653
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.