Learn More
Medchemexpress LLC DBCO-acid | 1353016-70-2 | 99.7% | 305.33 | C19H15NO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HY429725G
Encompass_Preferred
DBCO-acid is a dibenzocyclooctyne-containing carboxylic acid used as a click chemistry reagent for strain-promoted alkyne-azide cycloaddition (SPAAC). It is commonly used in bioconjugation to attach payloads to azide-functionalized molecules and to prepare activated esters for linker and linker-payload synthesis.
  • Enables copper-free click conjugation (SPAAC).
  • Used to prepare activated esters and linker conjugates.
  • High purity suitable for research and synthesis.
  • Available in multiple pack sizes, including 5 g.
Catalog No. 50-002-02916