Learn More
Broadpharm DBCO-PEG4-NHS ester | 1427004-19-0 | MFCD26793797 | 25 mg
Supplier:  Broadpharm BP2228825MG
Encompass
DBCO-PEG4-NHS ester | Purity: 97% | Mol Wt: 649.7 | 1427004-19-0 | MFCD26793797 | 25 mg
Catalog No. NC1548969
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.