Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DBE dibasic ester

Catalog No. 4220531L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-205-31L 1 L
42-205-34L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-205-31L Supplier Sigma Aldrich Supplier No. 4220531L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mixture of dimethyl glutarate, dimethyl succinate and dimethyl adipate

Specifications

Density 1.092 g/mL (at 25°C)
Boiling Point 196°C to 225°C
Refractive Index n20/D 1.424 (literature)
Linear Formula CH3O2C(CH2)nCO2CH3 (n=2,3,4)
Quantity 1 L
MDL Number MFCD00152995
Synonym DBE; Dibasic ester mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.