Learn More
3M™ DBI-SALA™ Advanced™ Floor Mount Cast-in-Place Sleeve Davit Base
Catalog No. 19809732 Shop All 3M Products
Change view
Click to view available options
Silver
Catalog No.
19-809-732 Silver
1 options
Catalog No. 19-809-732 Supplier 3M™ Supplier No. 7100311211
Add to Cart
Add to Cart
3M™ DBI-SALA™ Advanced™ Floor Mount Cast-in-Place Sleeve Davit Base is designed for manhole and confined space entry/retrieval applications.
  • Floor mount cast-in-place sleeve davit base
  • Built-in pins for fresh concrete
  • Detent style attachment
  • Flush mount design
  • Highly corrosion resistant stainless steel hardware
  • High strength stainless steel construction
  • 360 degree davit mast rotation
  • PVC davit mast sleeve liner
  • Permanent installation with standard tools
  • Also available in zinc plated steel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Mount Cast-in-Place Sleeve Davit Base
Weight (Metric) 9.71 kg
Weight (English) 21.4 lb.
Dimensions (L x W x H) 30.4 x 20.3 x 23.6 cm