Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Advanced™ Portable Fall Arrest Post

Catalog No. 19070816 Shop All 3M Products
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-070-816 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-816 Supplier 3M™ Supplier No. 7100188842
  • $8,900.00
    $6,521.65 / Case of 1

    Save $2,378.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Portable, telescoping fall arrest post with three independent 360 degree swiveling anchor points, provides protection and complete mobility for three workers on top of transformers or other types of vertical platforms with potential fall hazards.

  • 360 degrees of protection for 3 workers
  • Telescoping design
  • Built-in levelling system
  • Anchor ring for horizontal lifeline system
  • Integral carrying handle
  • Lightweight, corrosion resistant aluminum construction
  • Order base, lanyard or SRL separately
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Aluminum
Weight (English) 40 lb.
Weight (Metric) 18 kg
Load Bearing Capacity (English) 310 lb.
Load Bearing Capacity (Metric) 140 kg
Style 360 degrees of protection for 3 workers
Product Type Portable install
For Use With (Equipment) Order base, lanyard or SRL separately
Certifications/Compliance OSHA 1910.66, OSHA 1926.502
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.