Learn More
3M™ DBI-SALA™ Advanced™ Vehicle Hitch Mount Universal Joint Assembly
Catalog No. 19809728 Shop All 3M Products
Change view
Click to view available options
Silver
Catalog No.
19-809-728 Silver
1 options
Catalog No. 19-809-728 Supplier 3M™ Supplier No. 7012820550
  • $2,717.00
    $1,991.65 / Case of 1
    Save $725.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Universal joint assembly for vehicle hitch mount sleeve and extension.
  • Installs between vehicle hitch mount sleeve and/or extension
  • Compensates for uneven ground conditions
  • Simple nut and bolt attachment
  • Durable and corrosion resistant construction
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vehicle Hitch Mount Universal Joint Assembly
Weight (English) 9.92 kg
Dimensions (L x W x H) 15 x 2 x 2 in.
Weight (Metric) 4.5 kg