Learn More
3M™ DBI-SALA™ Center Mount Sleeve Davit Base
Catalog No. 19809733 Shop All 3M Products
Change view
Click to view available options
Yellow zinc
Catalog No.
19-809-733 Yellow zinc
1 options
Catalog No. 19-809-733 Supplier 3M™ Supplier No. 7100230116
  • $2,073.00
    $1,518.65 / Case of 1
    Save $554.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
3M™ DBI-SALA™ Center Mount Sleeve Davit Base is designed for manhole and confined space entry/retrieval applications.
  • Centre mount sleeve davit base
  • Bolt to steel and concrete floors
  • Corrosion resistant plated hardware
  • High strength steel construction
  • 360 degree davit mast rotation
  • PVC davit mast sleeve liner
  • Permanent installation with standard tools
  • Also available in stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Center Mount Sleeve Davit Base
Weight (English) 23 lb.
Weight (Metric) 10.43 kg