Learn More
3M™ DBI-SALA™ Confined Space Floor Mount Sleeve Davit Base
Catalog No. 19809703 Shop All 3M Products
Change view
Click to view available options
Yellow, zinc
Catalog No.
19-809-703 Yellow, zinc
1 options
Catalog No. 19-809-703 Supplier 3M™ Supplier No. 7100235312
Add to Cart
Add to Cart
Floor mount sleeve davit base, flush mount design, zinc plated, for the offset davit mast.
  • Floor mount sleeve davit base
  • Bolt-in install to existing concrete or steel floors
  • Flush mount design with recessed fastener pockets
  • Corrosion resistant plated hardware
  • High strength steel construction
  • 360 degree davit mast rotation
  • PVC davit mast sleeve liner
  • Permanent installation with standard tools
  • Also available in stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Confined Space Tripod Mounting Base
Dimensions (L x W x H) 30.4 x 20.3 x 23.6 cm
Weight (Metric) 7.62 kg
Weight (English) 16.8 lb.