Learn More
3M™ DBI-SALA™ Delta™ Cross-Over Style Positioning and Climbing Harness
Catalog No. 19809226 Shop All 3M Products
Change view
Click to view available options
Size:
X-Small
Catalog No. Size
19-809-226 X-Small
1 options
Catalog No. 19-809-226 Supplier 3M™ Supplier No. 1103253
Back, front and side D-rings, pass-thru buckle leg straps.
  • No-tangle design
  • Stand-up back D-ring
  • Front D-ring
  • Side D-rings
  • Pass-thru buckle leg straps
  • Repel™ webbing
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material Repel Polyester
D-Ring Location Back, Front, Side
Certifications/Compliance ANSI A10.32, ANSI Z359.4, ANSI Z359.1, OSHA 1910.66, OSHA 1926.502, ANSI Z359.3
Webbing Material Repel™ Polyester
Style Full-body vest
Product Type Cross-Over Style Positioning, Climbing Harness
Weight (English) 3.8 lb.
Size X-Small