Learn More
3M™ DBI-Sala™ Crossover Full-Body Harnesses
Catalog No. 19066811 Shop All 3M Products
Change view
Click to view available options
D-Ring Location:
Back, Front, Side
Front, Side
Side
Without side
Size:
Small, Medium, Large
Universal
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. D-Ring Location Size
19-066-811 Front, Side Universal
19-066-810 Back, Front, Side Universal
18-705 Side Small, Medium, Large
18-708 Without side Small, Medium, Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-066-811 Supplier 3M™ Supplier No. 7012151389
Add to Cart
Add to Cart
Lightweight polyester for all-day wear
  • Feature patented crossover design with adjustable front D-ring, used for positioning or ladder climbing
  • Back D-ring is used for fall arrest or restraint
  • Friction buckles on shoulders and mating pass-through adjuster buckles on lower shoulder straps and leg straps
  • Side D-rings for additional restraint or positioning options
  • Suitable for fall arrest, restraint, or positioning
Compliance
Meet ANSI Z359.1-1992, ANSI A10.14-1991, and OSHA requirements.
TRUSTED_SUSTAINABILITY
Specifications
Material Lightweight Polyester
Size Universal
D-Ring Location Front, Side