Learn More
3M™ DBI-SALA™ Delta™ II No-Tangle™ Construction Harnesses
Catalog No. 19809221 Shop All 3M Products
Change view
Click to view available options
Size:
2X-Large
Large; X-Large tongue buckle body belt, leg straps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-809-221 2X-Large
19-822-115 Large; X-Large tongue buckle body belt, leg straps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-809-221 Supplier 3M™ Supplier No. 7012815358
Add to Cart
Add to Cart
No-tangle design holds harness shape for easy donning
  • Body belt and side D-rings
  • Lightweight polyester webbing
  • Plated forged alloy steel hardware
  • Unique spring-loaded stand-up dorsal D-ring with impact indicator
  • Five-point adjustment
  • Nonslip, pass-thru buckle chest strap
Compliance
Meet or exceed ANSI™ Z359 and applicable OSHA regulations.
TRUSTED_SUSTAINABILITY
Specifications
Material Polyester Webbing
D-Ring Location Back, Side
Size 2X-Large
Certifications/Compliance Meets or exceeds ANSI Z359 and applicable OSHA regulations.