Description
- Spring-loaded stand-up dorsal D-ring design ensures fast, easy and safe connections
- Rip-stitch indicators allow to inspect the harness at glance, for prior damaging impact loads
- Harness falls into place, allowing quick and easy donning
- Side D-rings for positioning
- Adjustable non-slip chest strap with easy to use pass-thru buckle
- Tongue buckle leg straps for ease of use and will not loose
- Lanyard keepers holds unused snap hooks and help reduce trip hazard
- Revolver vertical torso adjuster provides simple and fast adjustment eliminating loose ends and locks webbing into place preventing slippage
- Velcro cover protects harness labels and holds I-Safe™ ID tag
Compliance
Meets ANSI™ Z359.11 requirements.
Specifications
Specifications
| Material | Polyester Webbing |
| D-Ring Location | Back |
| Size | Small |
| Buckle Style (Chest) | Pass-Through |
| Buckle Style (Leg) | Tongue Buckle |
| Style | Vest Style |