Learn More
3M™ DBI-SALA™ Delta™ Vest-Style Retrieval Harness
Catalog No. 19809722 Shop All 3M Products
Change view
Click to view available options
Material:
Zinc-plated Steel, Tech-Lite Aluminum
Catalog No. Material
19-809-722 Zinc-plated Steel, Tech-Lite Aluminum
1 options
Catalog No. 19-809-722 Supplier 3M™ Supplier No. 7100262935
Add to Cart
Add to Cart
3M™ DBI-SALA™ Delta™ Vest-Style Retrieval Harness features back and shoulder D-rings and quick connect buckle leg straps
  • No-tangle design
  • Stand-up back D-ring
  • Shoulder D-rings
  • Lightweight aluminum quick connect buckle leg straps
  • Repel™ webbing
  • Built-in lanyard keepers
  • Revolving vertical torso buckle
  • Impact Indicator
  • Protected labels, equipped with 3M™ Connected Safety ID (CSID)
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material Zinc-plated Steel, Tech-Lite Aluminum
Product Type Vest-Style Retrieval Harness
Size Universal
Webbing Material Repel™ Polyester
Capacity (English) 420 lb.
Weight (English) 3.8 lb.