Learn More
3M™ DBI-SALA™ Delta™ Vest-Style Retrieval Harness
Catalog No. 19809198 Shop All 3M Products
Change view
Click to view available options
Size:
2X-Large
3X-Large
Universal
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-809-198 X-Large
19-809-201 3X-Large
19-999-103 2X-Large
18-823 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-809-198 Supplier 3M™ Supplier No. 7100157104
Add to Cart
Add to Cart
This item is not returnable. View return policy
Back and shoulder D-rings, tongue buckle leg straps.
  • No-tangle design
  • Stand-up back D-ring
  • Shoulder D-rings
  • Tongue buckle leg straps
  • Repel™ webbing
  • Built-in lanyard keepers
TRUSTED_SUSTAINABILITY
Specifications
Webbing Material Repel™ Polyester
Style Vest
D-Ring Location Back, Shoulder
Size X-Large
Certifications/Compliance OSHA 1926.502, ANSI Z359.1, ANSI A10.32, ANSI Z359.4, OSHA 1910.66
Weight (English) 4.2 lb.
Product Type Vest-Style Retrieval Harness