Learn More
3M™ DBI-SALA™ Delta™ Vest-Style Retrieval Harness
Catalog No. 19999103 Shop All 3M Products
Change view
Click to view available options
Size:
2X-Large
3X-Large
Universal
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-103 2X-Large
19-809-198 X-Large
19-809-201 3X-Large
18-823 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-999-103 Supplier 3M™ Supplier No. 7012815216
Add to Cart
Add to Cart
Back and shoulder D-rings, tongue buckle leg straps.
  • No-tangle design
  • Stand-up back D-ring
  • Shoulder D-rings
  • Tongue buckle leg straps
  • Repel™ webbing
  • Built-in lanyard keepers
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets or exceeds applicable standards including OSHA/ANSI/ANSIFor Use With (Application) Z359
Size 2X-Large
Style Vest
Webbing Material Repel™ Polyester
Weight (English) 4.2 lb.
Product Type Vest-Style Retrieval Harness
D-Ring Location Back, Shoulder