Learn More
3M™ DBI-SALA™ ExoFit STRATA™ Vest-Style Harness
Catalog No. 19040602 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-040-602 Large
19-040-603 X-Large
19-040-621 Small
19-040-622 Medium
19-040-623 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-040-602 Supplier 3M™ Supplier No. 7012815926
Add to Cart
Add to Cart
Light-weight fall protection harness designed to help elevate employees safety and comfort.
  • PolarMesh™ back pad
  • EZ-Link™ SRL adapter
  • Tri-Lock Revolver™ quick connect buckles
  • Stand-up Tech-Lite™ aluminum dorsal D-ring
  • Repel™ webbing
  • Built-in lanyard keepers that automatically reset
  • Revolver™ vertical torso adjusters
  • Impact Indicator
  • Integrated suspension trauma straps
  • Customizable name tag
  • Protected labels, equipped with i-Safe™
  • Fall protection for tools attachment point
TRUSTED_SUSTAINABILITY
Specifications
D-Ring Location Dorsal
Buckle Style Tri-Lock Revolver™ Quick Connect
Weight (English) 5.35 lb.
Size Large
Certifications/Compliance ANSI Z359.1, ANSI Z359.11, OSHA 1910.66, OSHA 1926.502
Color Gray