Learn More
3M™ DBI-SALA™ Lad-Saf™ Grab Bar Extension Bracket
Catalog No. 19809403 Shop All 3M Products
Change view
Click to view available options
Product Type:
Grab Bar Extension Top Bracket for Fixed Ladder
Catalog No. Product Type
19-809-403 Grab Bar Extension Top Bracket for Fixed Ladder
1 options
Catalog No. 19-809-403 Supplier 3M™ Supplier No. 7100311822
  • $973.00
    $713.65 / Each of 1
    Save $259.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Grab bar extension for fixed ladder.
  • Grab bar extension for Lad-Saf™ fixed ladder safety system
  • Telescoping extension can be raised or lowered
  • User installed system is fast, easy and safe
  • Extremely rugged and corrosion resistant construction
  • Customizable to suit almost any application
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance FAA RR-S-001301, OSHA 1910.27, ANSI A14.3-1992
Dimensions (L x W) 3 x 2 in.
Dimensions (L x W x H) 3 x 2 x 49.5 in.
Product Type Grab Bar Extension Top Bracket for Fixed Ladder
For Use With (Equipment) Fixed Ladders,Slides into 6116054 top bracket providing egress tie-off point
Weight (English) 18 lb.