Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Medium Duty Tool Cinch

3M Company DBI-SALA™ Medium Duty Tool Cinch features a unique cinch design that allows users to quickly secure even the most difficult to tether tools, and continue their work

$371.92 - $474.33

Specifications

For Use With (Application) Tethering and securing tools
Load Bearing Capacity (English) 35 lb.
Load Bearing Capacity (Metric) 15.9 kg
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
19-070-721
View Documents
3M™
7012689168
Dual Wing Medium Duty Tool Cinch
Pack of 10 for $474.33
Only null left
Add to Cart
 
19-070-764
View Documents
3M™
7100310175
Single Wing Medium Duty Tool Cinch
Pack of 10 for $371.92
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Tethering and securing tools
35 lb.
15.9 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.