Learn More
3M™ DBI-SALA™ Personal Self-Rescue Kit, Rope
Catalog No. 17018260 Shop All 3M Products
Change view
Click to view available options
Size:
100 ft.
Catalog No. Size
17-018-260 100 ft.
1 options
Catalog No. 17-018-260 Supplier 3M™ Supplier No. 7100307007
Add to Cart
Add to Cart
Self-Rescue Device for harnesses provides crew members the ability to rescue themselves in challenging situations. This rescue pack contains rope and easily accessible D-ring This rescue device has a universal design that integrates with most harnesses, making it a premium piece of safety gear.
  • Premium detachable safety kit lets them prepare for the worst with a strong lifeline
  • Easily accessible D-ring for secure tethering, and an assisted-rescue ring if workers are injured or semi-conscious.
  • Lightweight emergency harness device is built to be small and streamlined so that it doesn't add excessive weight, cause distractions, or restrict a worker's movement.
  • Tough nylon pack attaches to any full-body harness with its universal design, and it contains durable rope within a streamlined compartment.
  • The personal descent safety product resists abrasions, weather, and wear and tear, providing peace of mind when working up high.
  • Easy-to-use rescue equipment is made with molded padding, providing durable comfort for workers suspended in mid-air.
  • Its lightweight, compact construction makes it easy to carry, store, and wear without a second thought.
TRUSTED_SUSTAINABILITY
Specifications
Size 100 ft.
Product Type Self-Rescue Device