Learn More
3M™ DBI-SALA™ Bosun Chair with Belt and Support Straps
Catalog No. 19823746 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-823-746 Small
19-809-181 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-823-746 Supplier 3M™ Supplier No. 7012815067
  • $1,056.00
    $773.65 / Each of 1
    Save $282.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Workseat with 7.75 in. x 18 in. x .75 in. (19.7 x 45.7 x 1.9 cm) rigid board, suspension D-rings, body belt with pad and back D-ring, adjustable leg and shoulder straps.
  • Workseat with 7.75 in. x 18 in. x .75 in. (19.7 x 45.7 x 1.9 cm) rigid board
  • Built-in dual suspension D-rings
  • Integrated tongue buckle body belt and hip pad
  • Back D-ring for tagline or tools
  • Adjustable leg and shoulder straps
  • Durable polyester construction
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material Latex-treated Polyester
Includes Seat board, tongue buckle body belt, 3 in. pad, back and suspension D-ring, adjustable leg and shoulder straps
Size Small
Certifications/Compliance OSHA 1910.66, OSHA 1926.502
Product Type Bosun Chair with Belt and Support Straps
Weight (English) 8 lb.