Learn More
3M™ DBI-Sala™ Vertical Rope Lifelines
Self locking snap hook at one end and tapered at other for easy connection to anchorage
Supplier: 3M™ 7100262938
Description
- Polyester/polypropylene blend rope for strength and durability
Compliance
Meets or exceeds all applicable industry standards including OSHA, ANSI and the stringent ANSI Z359.
Specifications
7.62 m | |
25 ft. | |
1.6 cm | |
0.63 in. |
Lifeline Rope | |
OSHA, ANSI, ANSI Z359 | |
Self locking snap hook at one end and tapered at other |