Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DBM 1285 dihydrochloride
GREENER_CHOICE

Catalog No. p-5545765
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 50-951-0R

Supplier: Tocris Bioscience™ 5095/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

p38 MAPK inhibitor; anti-inflammatory

DBM 1285 dihydrochloride is a p38 MAPK inhibitor. Supresses p38 phosphorylation and LPS-induced TNF-α production in macrophages and in vivo. Attenuates zymosan-induced inflammation and adjuvant-induced arthritis in murine models. Orally active.

Specifications

Quantity 10 mg
Product Type DBM 1285 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 468.42
Inhibitors DBM 1285 dihydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.