Learn More
Apexbio Technology LLC WZ4002 1213269-23-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A13895.1
Encompass_Preferred
WZ4002 (CAS 1213269-23-8) is an irreversible EGFR tyrosine kinase inhibitor that selectively targets mutant epidermal growth factor receptor (EGFR) variants It potently inhibits autophosphorylation of EGFR mutants including T790M L858R/T790M delE746 A750/T790M L858R and delE746 A750 at low nanomolar Ki values Compared to wild-type EGFR mutant EGFR demonstrates greater sensitivity to WZ4002 at lower concentrations limiting off-target toxicity in normal tissues WZ4002 serves as a research tool for studying EGFR-driven tumorigenesis and therapeutic resistance in oncology models
Catalog No. 50-000-63014
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.