Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DBR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100095 Supplier Invitrogen™ Supplier No. PA5100095
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DBR1.

The RNA lariat debranching enzyme, or DBR1, specifically hydrolyzes 2-prime-to-5-prime branched phosphodiester bonds at the branch point of excised lariat intron RNA and converts them into linear molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DBR1
Gene Accession No. Q923B1, Q9UK59
Gene Alias AW018415; Dbr1; debranching enzyme homolog 1; debranching enzyme homolog 1 (S. cerevisiae); debranching RNA lariats 1; lariat debranching enzyme; RNA lariat debranching enzyme
Gene Symbols DBR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DBR1(Accession Q9UK59), corresponding to amino acid residues Y106-I156.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51163, 681234, 83703
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.