Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DC 271

Catalog No. 68735
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-735 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 68-735 Supplier Tocris Bioscience™ Supplier No. 6873/5
This item is not returnable. View return policy

Fluorescent retinoic acid analog; solvochromatic probe

DC 271 is a fluorescent retinoic acid analog (retinoid all-trans-retinoic acid; ATRA) (Kd = 42 nM at cellular retinoid binding protein II (CRABPII)). Solvochromatic probe: fluorescent λex, λem, and quantum yield affected by solvent type. Activates same genes as endogenous retinoic acid in human keratinocyte cells (HaCaT) when incubated for 72 h at 1 μM. Also elicits cellular responses consistent with those of retinoic acid. Excitation λ = 351, 350, 382 and 378 nm in DMSO, EtOH, DCM and Toluene, respectively; emission λ = 442, 461, 572 and 447 nm in DMSO, EtOH, DCM and Toluene, respectively. Quantum yield = 19.3, 1.14, 71.45, 80.21 % in DMSO, EtOH, DCM and Toluene, respectively

Specifications

Chemical Name or Material 4-[2-[1,2,3,4-Tetrahydro-4,4-dimethyl-1-(1-methylethyl)-6-quinolinyl]ethynyl]benzoic acid
CAS 198696-03-6
Quantity 5 mg
Target Fluorescent Receptor Probes
Molecular Formula C23H25NO2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.