Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific DC Ammeter, 0-1A/0-5A
SDPSCI_ED

Catalog No. S81078 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Electricity Demonstration
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S81078 Electricity Demonstration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81078 Supplier United Scientific Supplier No. MAM002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammeter for use in electricity demonstrations.

  • Mounted in a black plastic case
  • Meters have a scale length of 75 mm
  • ± 2.5% accuracy and comes with 5-way binding post and external calibrating screw
  • Meters are not protected against prolonged or significant overload
  • Stackable for storage
  • Prop 65 Warning Label

Specifications

For Use With (Application) Electricity Demonstration
Product Type Ammeter
Sufficient For 0-1 A/0-5 A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.