Learn More
United Scientific™ DC Motor Kit 12 Pack
SDPSCI_ED
Catalog No. S000853 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W x H):
3 x 2.5 x 2 in.
Catalog No. Dimensions (L x W x H)
S000853 3 x 2.5 x 2 in.
1 options
Prop 65
Catalog No. S000853 Supplier United Scientific™ Supplier No. MOTOR12
Add to Cart
Add to Cart
Prop 65
This DC Motor Kit 12 Pack can be used to build a real working motor that does not require a magnet.
  • Learn the functions of each part of a motor, from the armature to the wire coils.
  • Simply add a battery and watch it spin.
  • Includes a step-by-step guide with assembly tips.
  • Each motor requires one AA battery (sold separately).
  • Measures 3 x 2.5 x 2 in. (7.6 x 6.3 x 5 )
  • Hands-on activity to build a real working motor
  • Shows how a magnet is not always required to build a DC motor
  • Includes activity guide
Required but not included: Wire strippers, sandpaper, small screw driver, needle nose pliers, ruler, AA battery.
Specifications
Product Type Motor Set
Dimensions (L x W x H) 3 x 2.5 x 2 in.