Learn More
Description
MA1-40070 detects SIGN-R1 from mouse samples. MA1-40070 has been successfully used in flow cytometry, functional assays, immunohistochemistry (paraffin) and immunohistochemistry (frozen) applications. The MA1-40070 immunogen is mouse SIGN-R1.
Specifications
Specifications
| Antigen | DC-SIGN |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | ER-TR9 |
| Concentration | Conc. not determined |
| Conjugate | Unconjugated |
| Formulation | Cell culture media with 0.02% sodium azide |
| Gene | Cd209a |
| Gene Accession No. | Q91ZX1 |
| Gene Alias | CD209; CD209 antigen; CD209 antigen-like protein A; CD209 molecule; Cd209a; CD209a antigen; CD209a molecule; Cd209d; CD209d antigen; CDSIGN; Cire; CLEC4L; Clec4m; C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; C-type lectin domain family 4, member M; Dcsign; DC-SIGN; DC-SIGN1; Dc-signr; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin; dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443; RGD1561104; SIGN-R1; SIGNR5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
